missing translation for 'onlineSavingsMsg'
Learn More

Cesium chloride, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog Number 11335009
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
7647-17-8
ClCs
168.36
MFCD00010955
AIYUHDOJVYHVIT-UHFFFAOYSA-M
cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride
24293
CHEBI:63039
caesium(1+) chloride
[Cl-].[Cs+]
This item is not returnable. View return policy
7647-17-8
ClCs
168.36
MFCD00010955
AIYUHDOJVYHVIT-UHFFFAOYSA-M
cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride
24293
CHEBI:63039
caesium(1+) chloride
[Cl-].[Cs+]

Cesium chloride is widely utilized as a precursor for the source of cesium ions. It finds an application in analytical chemistry and acts as a reagent to identify inorganic ions like aluminum, gallium and chromium by the color and morphology of the precipitate. Its radioisotopes are employed in nuclear medicine applications such as treatment of cancer and diagnosis of myocardial infarction. Occasionally, it can act as a phase transfer catalyst in selected organic reactions. It is applied in the manufacturing of electrically conducting glasses and screens of cathode ray tubes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 1303°C
Melting Point 646°C
Color White
Physical Form Crystalline
Quantity 10 g
Assay Percent Range 99.99%
Solubility Information Soluble in water and alcohol. Insoluble in acetone.
Formula Weight 168.36
Odor Odorless
Refractive Index 1.6418
Sensitivity Hygroscopic
Appearance White powder/crystalline/beads
Density 3.99 g/cm3 at 20°C
Chemical Name or Material Cesium chloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'