Learn More
Cesium chloride, 99+%, pure, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.

Specifications
Specifications
Boiling Point | 1290.0°C |
Melting Point | 646.0°C |
Color | White |
Physical Form | Crystals |
Quantity | 100 g |
Assay Percent Range | 99% min. (Argentometry) |
Linear Formula | CsCl |
Merck Index | 15,201 |
Solubility Information | Solubility in water: 1860g/L (20°C). Other solubilities: soluble in alcohol,insoluble in acetone |
Formula Weight | 168.36 |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0464350 | Certificate of Analysis | 17/07/2024 | 422850250,422855000,422851000 |
A0461103 | Certificate of Analysis | 15/03/2024 | 422850250,422855000,422851000 |
A0445889 | Certificate of Analysis | 14/09/2023 | 422850250,422855000,422851000 |
A0398884 | Certificate of Analysis | 14/12/2022 | 422850250,422855000,422851000 |
A0395714 | Certificate of Analysis | 14/12/2022 | 422850250,422855000,422851000 |
Safety and Handling

- Cesium chloride
- Warning
- Reproductive toxicity Category 2
- H361-Suspected of damaging fertility or the unborn child.
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.