missing translation for 'onlineSavingsMsg'
Learn More
Cesium Chloride, MP Biomedicals™ Product Code.: 11401662

Cesium Chloride, MP Biomedicals™

Product Code. 11401662 Shop All MP Biomedicals Products
1 kg
Click to view available options
Quantity:
100 g
500 g
1 kg
This item is not returnable. View return policy

Product Code. 11401662

Brand: MP Biomedicals 04813069

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cesium Chloride Ultra Pure, 99.999%, White Powder

  • Cesium chloride is typically used for density gradient work and for the purification of virus/phage, nucleic acids and nucleoproteins.
  • It is used for the preparation of electrically conducting glasses, used to make solutions for the separation of RNA from DNA by density gradient centrifugation.
  • Cesium chloride (CsCl) is commonly used in the separation of DNA and RNA.
  • Cesium chloride , an inorganic compound, is an important source of cesium ions in a variety of applications.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-17-8
Molecular Formula ClCs
Molecular Weight (g/mol) 168.36
MDL Number MFCD00010955
InChI Key AIYUHDOJVYHVIT-UHFFFAOYSA-M
Synonym Cesium Monochloride, Dicesium Dichloride
IUPAC Name caesium(1+) chloride
SMILES [Cl-].[Cs+]

Specifications

Boiling Point 1303°C (literature)
Melting Point 646°C (literature)
Color White
pH 5.5 to 6.5 (1% aq soln)
Physical Form Powder
Quantity 1 kg
Solubility Information Freely soluble in water (500mg/mL),(50% aqueous-clear, colorless solution); soluble in ethanol, alcohol
Grade 0.99999
Refractive Index 1.642(Lit.)
Density 3.99g/mL(Lit.)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cesium Chloride, MP Biomedicals™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.