missing translation for 'onlineSavingsMsg'
Learn More
Cesium hydroxide hydrate, 99.9% (metals basis), Thermo Scientific Chemicals Product Code.: 11386678

Cesium hydroxide hydrate, 99.9% (metals basis), Thermo Scientific Chemicals

Product Code. 11386678
100 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11386678

Brand: Thermo Scientific Alfa Aesar 013233.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12260-45-6
Molecular Formula CsHO
Molecular Weight (g/mol) 149.91
MDL Number MFCD00010964
InChI Key HUCVOHYBFXVBRW-UHFFFAOYSA-M
Synonym cesium hydroxide monohydrate, cesium hydroxide hydrate, cesium hydroxide, monohydrate, cesium hydroxide-hydrate, cesium hydroxide, hydrate, ksc493c5l, caesium 1+ ion hydrate hydroxide, cesium hydroxide,monohydrate 9ciShow MoreShow Less
PubChem CID 23679066
SMILES [OH-].[Cs+]

Specifications

Color White
Physical Form Crystalline
Quantity 100 g
Assay Percent Range 99.9% (Metal basis)
Solubility Information Soluble in water and ethanol.
IUPAC Name cesium;hydroxide;hydrate
Formula Weight 149.91 (Anhydrous)
Odor Odorless
Sensitivity Hygroscopic
Appearance White crystalline
Chemical Name or Material Cesium hydroxide hydrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberX24K021 Certificate TypeCertificate of Analysis Date08/11/2024 Catalog Number013233.06,013233.14,013233.22,013233.A1
Lot NumberY15H060 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number013233.06,013233.14,013233.22
Lot NumberB06U010 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number013233.06,013233.14,013233.22
Lot NumberA29P20 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number013233.06,013233.14,013233.22
Lot NumberH27Y031 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number013233.06,013233.14,013233.22
compliance-icons compliance-icons
Product Identifier
  • Caesium hydroxide hydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Caesium hydroxide hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cesium hydroxide hydrate, 99.9% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.