missing translation for 'onlineSavingsMsg'
Learn More
Cesium sulfate, 99+%, pure, Thermo Scientific Chemicals Product Code.: 10703031

Cesium sulfate, 99+%, pure, Thermo Scientific Chemicals

Product Code. 10703031
25 g, Glass bottle
Click to view available options
Quantity:
10 g
25 g
100 g
250 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10703031

Brand: Thermo Scientific Acros 218700250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10294-54-9
Molecular Formula Cs2O4S
Molecular Weight (g/mol) 361.87
MDL Number MFCD00010959
InChI Key FLJPGEWQYJVDPF-UHFFFAOYSA-L
Synonym cesium sulfate, dicesium sulfate, sulfuric acid, dicesium salt, caesium sulphate, caesium sulfate, unii-8d6r91cs62, dicaesium 1+ ion sulfate, cesiumsulfate, dicaesium 1+ sulfateShow MoreShow Less
PubChem CID 25137
SMILES [Cs+].[Cs+].[O-]S([O-])(=O)=O

Specifications

Boiling Point 1900.0°C
Melting Point 1005.0°C
Color White
Physical Form Fine Crystalline Powder
Quantity 25 g
Assay Percent Range 99% min
Linear Formula Cs2SO4
Merck Index 15,2015
Solubility Information Solubility in water: 1820g/L (25°C). Other solubilities: practically insoluble in alcohol, aceton, pyridine
IUPAC Name dicaesium(1+) sulfate
Formula Weight 361.86
Percent Purity ≥99%
Grade Pure
Packaging Glass bottle
Density 4.24
Chemical Name or Material Cesium sulfate, 99+%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2375462 Certificate TypeCertificate of Analysis Date21/02/2025 Catalog Number11520586,21870084,21870076,218700250,218701000,218702500
Lot Number2537758 Certificate TypeCertificate of Analysis Date21/02/2025 Catalog Number11520586,21870084,21870076,218700250,218701000,218702500
Lot Number2392917 Certificate TypeCertificate of Analysis Date21/02/2025 Catalog Number11520586,21870084,21870076,218700250,218701000,218702500
Lot Number2517121 Certificate TypeCertificate of Analysis Date21/02/2025 Catalog Number11520586,21870084,21870076,218700250,218701000,218702500
Lot Number2193222 Certificate TypeCertificate of Analysis Date21/02/2025 Catalog Number11520586,21870084,21870076,218700250,218701000,218702500

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cesium sulfate, 99+%, pure, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.