missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cesium sulfate, 99+%, pure, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
25 g
100 g
250 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Boiling Point | 1900.0°C |
Melting Point | 1005.0°C |
Color | White |
Physical Form | Fine Crystalline Powder |
Quantity | 250 g |
Assay Percent Range | 99% min |
Linear Formula | Cs2SO4 |
Merck Index | 15,2015 |
Solubility Information | Solubility in water: 1820g/L (25°C). Other solubilities: practically insoluble in alcohol, aceton, pyridine |
IUPAC Name | dicaesium(1+) sulfate |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 6
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2375462 | Certificate of Analysis | 21/02/2025 | 11520586,21870084,21870076,218700250,218701000,218702500 |
2537758 | Certificate of Analysis | 21/02/2025 | 11520586,21870084,21870076,218700250,218701000,218702500 |
2392917 | Certificate of Analysis | 21/02/2025 | 11520586,21870084,21870076,218700250,218701000,218702500 |
2517121 | Certificate of Analysis | 21/02/2025 | 11520586,21870084,21870076,218700250,218701000,218702500 |
2193222 | Certificate of Analysis | 21/02/2025 | 11520586,21870084,21870076,218700250,218701000,218702500 |
Safety and Handling


Product Identifier
- Cesium sulfate
- Warning
- Acute toxicity Category 4
- Reproductive toxicity Category 2
- Specific target organ toxicity after repeated exposure Category 2
- H302-Harmful if swallowed.
- H361f-Suspected of damaging fertility.
- H373-May cause damage to organs.
- P201-Obtain special instructions before use.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- MIXTURE LIST-Contains : Cesium sulfate
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cesium sulfate, 99+%, pure, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction