missing translation for 'onlineSavingsMsg'
Learn More

Chromacol Regenerated Cellulose Acetate Syringe Filters

Thermo Scientific™ Chromacol Regenerated Cellulose Acetate Syringe Filters are offered in a selection of diameters and pore sizes.

£131.95 - £236.00

Specifications

Type Syringe Filter
View More Specs

Products 5
Product Code Brand Material Diameter (Metric) Pore Size Price Quantity & Availability  
Product Code Brand Material Diameter (Metric) Pore Size Price Quantity & Availability  
12366878
View Documents Promotion Details
Thermo Scientific Chromacol
17-SF-02(RC)
Regenerated Cellulose 17 mm 0.2 μm
£236.00
Pack of 100
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
 
12334777
View Documents Promotion Details
Thermo Scientific Chromacol
30-SF-07(GMF)
Glass Fiber 30 mm 0.7 μm
£235.00
Pack of 100
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
 
12354777
View Documents Promotion Details
Thermo Scientific Chromacol
30-SF-12(GMF)
Glass Fiber 30 mm 1.2 μm
£156.00
Pack of 100
View alternative products
Save Up to 
»
Estimated Shipment: 08-08-2024
to see stock.
Add to basket
 
12336888
View Documents Promotion Details
Thermo Scientific Chromacol
17-SF-45(RC)
Regenerated Cellulose 17 mm 0.45 μm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12374797
View Documents Promotion Details
Thermo Scientific Chromacol
4-SF-02(RC)
Regenerated Cellulose 4 mm 0.2 μm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Thermo Scientific™ Chromacol Regenerated Cellulose Acetate Syringe Filters are offered in a selection of diameters and pore sizes.

Specifications

Specifications

Syringe Filter
Videos
SDS
Documents

Documents

Product Certifications