Learn More
Macherey-Nagel™ Chromafix™ SiOH Cartridge
Brand:  Macherey-Nagel™ 731830
 View more versions of this product
Product Code. 10023031
  • £177.00 / Pack of 50
Estimated Shipment: 03-03-2025
to see stock.
Explore more special offers
Add to basket
Specifications
Weakly Acidic Silica
Aflatoxins, Chloramphenicol, Pesticides, Steroids, Vitamins
SiOH
60 Å
500 m2/g
Adsorbent Weight: 880 mg
45 μm
Chromafix SiOH Cartridge
50 Pack
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Macherey-Nagel™ Chromafix™ SiOH Cartridge > Adsorbent Weight: 880mg