missing translation for 'onlineSavingsMsg'
Learn More
Chromium(III) oxide, 99.6% (metals basis), Thermo Scientific Chemicals Product Code.: 11363318

Chromium(III) oxide, 99.6% (metals basis), Thermo Scientific Chemicals

Product Code. 11363318
100 g
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy

Product Code. 11363318

Brand: Thermo Scientific Alfa Aesar 036258.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For vacuum deposition. It can be used in selective oxidation. It is mainly used for smelting metal chromium and chromium carbide. It is used as coloring agent for color painting on enamelware and porcelain ware, imitation leather or building material. It is also used for making sun-proof coating, abrasive, green polishing compound and special printing ink for printing bank note. It is used as catalyst for organic synthetic chromium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1308-38-9
Molecular Formula Cr2O3
Molecular Weight (g/mol) 151.99
MDL Number MFCD00010949
InChI Key UOUJSJZBMCDAEU-UHFFFAOYSA-N
Synonym chromium iii oxide, chromia, dichromium trioxide, chromium sesquioxide, chrome green, green chromium oxide, chromium 3+ oxide, green cinnabar, chrome ochre, chrome oxide
PubChem CID 517277
ChEBI CHEBI:48242
IUPAC Name dichromium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Cr+3].[Cr+3]

Specifications

Boiling Point 3000°C
Melting Point 2435°C
Physical Form Fused Lump
Quantity 100 g
Assay Percent Range 99.6% (metals basis)
Solubility Information Insoluble in water. insoluble in alcohol,acetone,acids.
Formula Weight 151.99
Refractive Index 2.551
Density 5.22 g/mL
Chemical Name or Material Chromium oxide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberY18K031 Certificate TypeCertificate of Analysis Date20/11/2024 Catalog Number036258.22,036258.14
Lot NumberZ12J063 Certificate TypeCertificate of Analysis Date08/01/2024 Catalog Number036258.22,036258.14
Lot NumberP18I011 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number036258.22,036258.14
Lot NumberN26H013 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number036258.22,036258.14
Lot NumberJ23L07 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number036258.22,036258.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Chromium(III) oxide, 99.6% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.