Learn More
Hirschmann™ Class AS AR™ Glass Volumetric Pipettes
Provides an extremely high degree of volumetric accuracy for your pipetting needs. Hirschmann Instruments™ Class AS AR™ Glass Volumetric Pipettes come with amber stain graduation marks.
Brand: Hirschmann™ 1340255
Description
- Acc. to DIN EN ISO 648
- Volume between two marks
- Conformity certified, with dated batch identification
Specifications
AS | |
Yes | |
6/Pk. | |
5 mL | |
Volumetric Pipet |
AR-GLAS | |
Brown | |
0.015 mL | |
410 mm |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.