Learn More
Sperian™ Honeywell™ Clearways™ CB20 Browguard with Ratchet Headband
Click to view available options
Weight (Metric):
94g
Description
- Recommended for the agriculture, chemical and forestry industries
- Protects against hazards including abrasion, airborne particles, biohazards, chemicals, impacts, splashes and more
- Visors available in polycarbonate
- **Face shield is not included**
Specifications
Specifications
Product Type | Browguard |
Weight (Metric) | 94g |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 12
Documents
Scientific Resources
Brochures
Applications and solutions series: Safety edition
File size: 7.7 MB
Product Content Correction
Product Title
Sperian™ Honeywell™ Clearways™ CB20 Browguard with Ratchet Headband >
Spot an opportunity for improvement?Share a Content Correction