Learn More
Nickel Electro™ Clifton™ 56 Litre Stainless Steel Flat Two Piece Lid with Insulated Handles
Two part stainless steel flat lids minimizing heat loss, saving energy, and assisting to maintain bath temperature.
Brand:  Nickel Electro™ LD-56
Product Code. 11875380
  • £235.00 / Each
Estimated Shipment: 20-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Two-part stainless steel flat lids minimize heat loss, save energy, and assist to maintain bath temperature
  • Fitted lid acts as a dust cover when Clifton water bath is not in use
  • Prevents contamination of liquid from airbourne particles
  • The lid is fitted with an insulated handle
Specifications
Specifications
Stainless Steel flat 2 Pc. lids with insulated handles
56 L Clfiton Unstirred Baths
Stainless Steel
2 kg
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Nickel Electro™ Clifton™ 56 Litre Stainless Steel Flat Two Piece Lid with Insulated Handles >