missing translation for 'onlineSavingsMsg'
Learn More
Cobalt(II) nitrate hexahydrate, 97.7% min, Thermo Scientific Chemicals Product Code.: 11341109

Cobalt(II) nitrate hexahydrate, 97.7% min, Thermo Scientific Chemicals

Product Code. 11341109
250 g
Click to view available options
Quantity:
25 g
250 g
1 kg
5 kg
This item is not returnable. View return policy

Product Code. 11341109

Brand: Thermo Scientific Alfa Aesar 011341.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cobalt(II) nitrate hexahydrate is used in the preparation of metallic cobalt and cobalt based dyes and pigments. It is a precursor for the preparation of catalyst for use in certain organic reactions. It finds use in catalysis, vitamin preparations, and porcelain decoration.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10026-22-9
Molecular Formula CoH12N2O12
Molecular Weight (g/mol) 291.03
MDL Number MFCD00149647
InChI Key QGUAJWGNOXCYJF-UHFFFAOYSA-N
Synonym cobalt nitrate hexahydrate, cobalt ii nitrate hexahydrate, cobaltous nitrate hexahydrate, cobalt dinitrate hexahydrate, cobalt 2+ nitrate hexahydrate, cobaltous nitrate, hexahydrate, kobalt ii-nitrat-hexahydrat, nitric acid, cobalt 2+ salt, hexahydrateShow MoreShow Less
PubChem CID 24821
ChEBI CHEBI:86214
IUPAC Name cobalt(2+);dinitrate;hexahydrate
SMILES O.O.O.O.O.O.[Co++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 55°C to 56°C
Color Red
Physical Form Crystalline
Quantity 250 g
UN Number UN1477
Merck Index 14,2444
Solubility Information Soluble in water,alcohol,acetone and ammonia.
Formula Weight 291.04 (182.94 Anhydrous)
Percent Purity ≤97.7%
Odor Odorless
Refractive Index 1.52
Density 1.87 g/mL
Chemical Name or Material Cobalt(II) nitrate hexahydrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberU16I025 Certificate TypeCertificate of Analysis Date23/08/2022 Catalog Number011341.30,011341.A1,011341.14,011341.A7
Lot NumberY16H016 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number011341.30,011341.A1,011341.14,011341.A7
Lot NumberY16H015 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number011341.30,011341.A1,011341.14,011341.A7
Lot NumberU24H014 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number011341.30,011341.A1,011341.14,011341.A7
Lot Number10208687 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number011341.30,011341.A1,011341.14,011341.A7

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cobalt(II) nitrate hexahydrate, 97.7% min, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.