missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cobalt(II) oxide, Co >74%, -325 mesh, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
100 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 1785.0°C |
Color | Brown-Green or Gray |
Physical Form | Powder |
Quantity | 10 g |
Assay Percent Range | 99+% |
Merck Index | 15,243 |
Solubility Information | Solubility in water: insoluble |
Formula Weight | 74.93 |
Percent Purity | >74% (Co) |
Packaging | Glass bottle |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0466009 | Certificate of Analysis | 10/09/2024 | 379201000,379200100 |
A0461745 | Certificate of Analysis | 12/06/2024 | 379201000,379200100 |
A0393419 | Certificate of Analysis | 14/12/2022 | 379201000,379200100 |
A0401248 | Certificate of Analysis | 14/12/2022 | 379201000,379200100 |
A0417584 | Certificate of Analysis | 20/10/2021 | 379201000,379200100 |
Safety and Handling



Product Identifier
- Cobalt(II) oxide
- Danger
- Acute toxicity Category 2
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H330-Fatal if inhaled.
- H301-Toxic if swallowed.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P311-Call a POISON CENTER/doctor/ .
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cobalt(II) oxide, Co >74%, -325 mesh, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction