missing translation for 'onlineSavingsMsg'
Learn More
Cobalt(II) oxide, Co >74%, -325 mesh, Thermo Scientific Chemicals Product Code.: 15159273

Cobalt(II) oxide, Co >74%, -325 mesh, Thermo Scientific Chemicals

Product Code. 15159273
100 g, Glass bottle
Click to view available options
Quantity:
10 g
100 g
Packaging:
Glass bottle
Este artículo no se puede devolver. Vea la política de devoluciones

Product Code. 15159273

Brand: Thermo Scientific Acros 379201000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Este artículo no se puede devolver. Vea la política de devoluciones

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 1307-96-6
Molecular Formula CoO
Molecular Weight (g/mol) 74.93
MDL Number MFCD00016031
InChI Key IVMYJDGYRUAWML-UHFFFAOYSA-N
Synonym cobalt ii oxide, zaffre, cobalt monoxide, cobalt black, cobalt oxide, cobaltous oxide, cobalt monooxide, monocobalt oxide, cobalt 2+ oxide, ci pigment black 13Show MoreShow Less
PubChem CID 14786
IUPAC Name oxocobalt
SMILES O=[Co]

Especificaciones

Melting Point 1785.0°C
Color Brown-Green or Gray
Physical Form Powder
Quantity 100 g
Assay Percent Range 99+%
Merck Index 15,243
Solubility Information Solubility in water: insoluble
Formula Weight 74.93
Percent Purity >74% (Co)
Packaging Glass bottle
Chemical Name or Material Cobalt(II) oxide
Mostrar más Mostrar menos

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0466009 Certificate TypeCertificate of Analysis Date10/09/2024 Catalog Number379201000,379200100
Lot NumberA0461745 Certificate TypeCertificate of Analysis Date12/06/2024 Catalog Number379201000,379200100
Lot NumberA0393419 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number379201000,379200100
Lot NumberA0401248 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number379201000,379200100
Lot NumberA0417584 Certificate TypeCertificate of Analysis Date20/10/2021 Catalog Number379201000,379200100
compliance-icons compliance-icons compliance-icons
Nombre del producto
  • Cobalt(II) oxide
Palabra de advertencia
  • Danger
Clasificación
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Indicaciones de peligro
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H330-Fatal if inhaled.
  • H301-Toxic if swallowed.
Consejos de prudencia
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P311-Call a POISON CENTER/doctor/ .

RUO – Research Use Only

Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto
Cobalt(II) oxide, Co >74%, -325 mesh, Thermo Scientific Chemicals >

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado