missing translation for 'onlineSavingsMsg'
Learn More

SI Analytics™ Conductivity Solution LF CSKC13

SI Analytics™ Conductivity Solution LF CSKC13 is designed as a test solution for measuring conductivity.

Brand:  SI Analytics™ 285126530

Product Code. 10234812

This item is not returnable. View return policy

Description

Description

  • Supplied in a PE bottle
  • Hermetically sealed in the glass ampoule and sterilized with hot steam, same as a pharmaceutical product
  • The ampoules can be easily opened at the breaking point
  • Tools are not required
  • Since refilling is not possible, you are always ensured of maximum calibration reliability
  • Maximum shelf life: unopened three months, opened six hours
TRUSTED_SUSTAINABILITY
Specifications

Specifications

250 mL
KCI 1.3 μS/cm
Conductivity measurements
25°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
SI Analytics™ Conductivity Solution LF CSKC13 > Capacity: 250 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.