missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Tri-Sil BP (BSA:Pyridine) Reagent

Employ a reagent-solvent mixture, for one-step derivatization with the Thermo Scientific™ Tri-Sil BP (BSA:Pyridine) Reagent,

Brand:  Thermo Scientific™ TS-49012

628.34 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 11567680

  • £633.15 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Employ a reagent-solvent mixture, for one-step derivatization with the Thermo Scientific™ Tri-Sil BP (BSA:Pyridine) Reagent, Composed of BSA and high-purity pyridine, this silylation reagent formulation derivatizes alcohols, phenols, organic acids, aromatic amides, and amines for GC analysis. It is excellent for use with unhindered steriods.

Reacts with a Wide Range of Compounds

Alcohols, phenols, some enols, and other hydroxyl/polyhydroxyl compounds to form trimethylsilyl ethers

Organic acids to form trimethylsilyl esters

Aromatic amides to form N-trimethylsilyl derivatives

Amines to form N-trimethylsilyl derivatives

Unhindered steroids

Not recommended for use with carbohydrates.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

GC
65 - 80, 20 - 25%
25 mL
Corrosive, Flammable
2°C to 8°C
Pyridine, Trimethylsilyl N-trimethylsilylacetamidate
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Tri-Sil BP (BSA:Pyridine) Reagent > 25mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.