Learn More
Conductivity Standard, NIST Traceable, 12,880μS/cm, Reagecon™
Shop All Reagecon Products

Description
This conductivity standard is aqueous based, thereby eliminating any errors attributable to matrix mismatch. Aqueous standards are non-hazardous for easy storage, handling and disposal. Each bottle is supplied with a comprehensive Certificate of Analysis and has a table of conductivity variation with temperature printed on the label.
- Convenience and lower costs—Reduced health, safety and environmental requirements and paperwork
- Very low temperature coefficient of variation—Reduces measurement errors and enables non-temperature controlled conditions or field work
- Guaranteed stability through expiry date—Eliminates the need to open a fresh bottle of standard every time it is used
- High accuracy (±1% at 25°C) —Enables it to be used as a calibrator or control, in accordance with the most exacting requirements (e.g. USP)
Specifications
Specifications
Quantity | 500 mL |
Certificate of Analysis | Yes |
Conductivity | 12,880μS/cm (±1% at 25°C) |
For Use With (Equipment) | Conductivity Meter |
Solvent or Matrix | Aqueous |
Product Suggestions
Customers who viewed this item also viewed
Documents
Scientific Resources
Brochures
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
CS1288025A1 | Certificate of Analysis | 26/03/2025 | CSKC12880 |
CS1288024J1 | Certificate of Analysis | 14/01/2025 | CSKC12880 |
CS1288024G1 | Certificate of Analysis | 07/08/2024 | CSKC12880 |
CS1288024A1 | Certificate of Analysis | 19/06/2024 | CSKC12880 |
CS1288023L1 | Certificate of Analysis | 22/04/2024 | CSKC12880 |
Special Offers
-
Multiple Manufacturers
Chemical Extravaganza
Shop now and save up to 97% on top-quality products for your lab needs.Promo Code: 25908Expires: 2025-03-31
Days
0Hours
7Minutes
32Seconds
32
Your input is important to us. Please complete this form to provide feedback related to the content on this product.