Learn More
YSI™ Conductivity/Temperature Cable
Brand:  YSI™ 60530-20
Product Code. 12706945
  • £464.00 / Each
Explore more special offers
Specifications
Conductivity/Temperature Cable
The measurement of conductivity, specific conductance, salinity, total dissolved solids and temperature, Professional Plus, Pro30
20 m
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
YSI™ Conductivity/Temperature Cable > Length Cable: 20m