Learn More
Corning™ BioCoat™ 96-Well, Fibronectin-Treated, Flat-Bottom Microplate
Color:Black, Clear Bottom; Sterility:Non-sterile; Lid:Without Lid
Brand: Corning™ 4584
Description
High optical quality, glass bottom, black microplates are ideal for performing high content cell-based assays using imaging systems. The glass bottom provides a flat and optically clear surface that reduces autofocus time, increases throughput, and is ideal for cell growth.
- High optical quality and scratch resistant glass
- Glass bottom thickness of 200 μm is well suited for imaging microscopy.
- Bottom flatness <50 μm to ensure planarity for imaging devices
- Low background fluorescence and minimal cross-talk provides the highest possible optical quality for cell-based assays.
- Half area 96-well microplate reduces reagent consumption.
Order Info
Supplied in ready-to-use Falcon™ Companion Tissue Culture Plates

Specifications
Half-Area | |
2°C to 8°C | |
8 x 12 | |
96 | |
Fibronectin | |
Promotion of cell attachment and spreading, rapid expansion of cell populations, increased proliferation of PC12 cells, studies of effects of collagen IV on cell behavior, cell adhesion assays | |
1/Pk. | |
Clear |
Without Lid | |
0.32 cm2 | |
Black, Clear Bottom | |
Non-sterile | |
Round, Flat Bottom (F-well) | |
10/Cs. | |
Human Fibronectin |
Product Suggestions
Customers who viewed this item also viewed
Your input is important to us. Please complete this form to provide feedback related to the content on this product.