missing translation for 'onlineSavingsMsg'
Learn More

Corning™ CellBIND™ Surface Cell Culture Flasks
GREENER_CHOICEBIOPHARMA

Increases suface wettability for more even and consistent cell attachment. Corning™ CellBIND™ Surface Cell Culture are available in a variety of sizes, designs and cap styles to meet all laboratory needs.

Brand:  Corning™ 3290

425.60 GBP valid until 2025-03-31
Use promo code "24073" to get your promotional price.


Product Code. 10327342

  • £448.00 / Pack of 100
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

  • Manufactured from optically clear, virgin polystyrene
  • Treated surface helps cells adapt to reduced serum or serum-free media; stable at room temperature
  • Ideal for short- and long-term cell culture
  • Available with vented caps containing a 0.2μm nonwettable membrane sealed to the cap, providing consistent, sterile gas exchange while minimizing risk of contamination; or phenolic-style caps for use in open systems requiring gas exchange
  • 3290 flask features redesigned neck for easier media pouring, lower height (1.4 in.) for reduced stacking height, and vented cap with Costar™-style concentric circle support grid geometry
  • Choice of canted or angled neck
    • Canted neck flasks allow easier pouring and improved access to the flask for pipetting or scraping
    • Angled neck improves pipetting access and reduces medium sloshing into the neck
  • Printed lot numbers provide easy traceability
  • Sterilized by gamma irradiation
  • Certified nonpyrogenic
  • 100% integrity tested

  • Flasks 431328 and 431346 are prelabeled with bar code for use with SelecT™ Robotic System

TRUSTED_SUSTAINABILITY
Specifications

Specifications

No
Orange
60 mL
For optimal cell attachment
Canted
Corning Cellbind
Sterile
2.02 oz.
HDPE
Polystyrene
Lot numbers for ease in traceability
Rectangular
Yes
Vented
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Information

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number21224021 Certificate TypeCertificate of Quality Date21/03/2025 Catalog Number3290
Lot Number20824020 Certificate TypeCertificate of Quality Date20/03/2025 Catalog Number3290
Lot Number20524018 Certificate TypeCertificate of Quality Date12/02/2025 Catalog Number3290
Lot Number18724014 Certificate TypeCertificate of Quality Date01/02/2025 Catalog Number3290
Lot Number14624011 Certificate TypeCertificate of Quality Date18/12/2024 Catalog Number3290
Special Offers

Special Offers

  • Corning Matrigel , Corning Biocoat , Corning CoolProduct , Corning Media Tech , Corning
    Corning Lab Products Offer
    Promo Code24073

    Expires2025-04-25

    New Offer
    Save Now
  • Multiple Manufacturers
    Browse exclusive prices for our academic customers
    Promo Code: 25081

    Expires2025-12-31

    New Offer
    Save Now
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Corning™ CellBIND™ Surface Cell Culture Flasks > Growth Area: 75cm2; Canted; Vent Cap; 100/cs

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.