missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Cell Culture Buffers: Dulbecco's Phosphate-Buffered Salt Solution 1X
BIOPHARMA

With low endotoxin content.

Brand:  Corning™ 21-030-CV

74.07 GBP valid until 2025-03-30
Use promo code "24134" to get your promotional price.


 View more versions of this product

Product Code. 15393571

  • £85.50 / Pack of 6

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Dulbecco's Phosphate Buffered Saline (DPBS) is a buffer solution which can be used to maintain cell culture media in the physiological pH range of 7.0 to 7.6. It can serve as an irrigating, transporting or diluting fluid while maintaining cell tonicity and viability for a limited period of time. This formulation contains calcium and magnesium.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Calcium and Magnesium
1 X
6 x 500 mL
6.9 to 7.1
2° to 30°C
DPBS
Calcium Chloride 0.10g/L, Potassium Chloride 0.2g/L, Potassium Dihydrogen Phosphate 0.2g/L, Magnesium Chloride Hexahydrate 0.1g/L, Sodium Chloride 8g/L, Disodium Phosphate Heptahydrate 2.1716g/L
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number30324007 Certificate TypeCertificate of Quality Date18/03/2025 Catalog Number21-030-CV
Lot Number25324006 Certificate TypeCertificate of Quality Date31/12/2024 Catalog Number21-030-CV
Lot Number21124009 Certificate TypeCertificate of Quality Date25/12/2024 Catalog Number21-030-CV
Lot Number27623017 Certificate TypeCertificate of Quality Date24/06/2024 Catalog Number21-030-CV
Lot Number07423001 Certificate TypeCertificate of Quality Date15/05/2024 Catalog Number21-030-CV
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Corning™ Cell Culture Buffers: Dulbecco's Phosphate-Buffered Salt Solution 1X > with Calcium and Magnesium; 36 Months; 500 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.