missing translation for 'onlineSavingsMsg'
Learn More
Cell Culture Grade Water Tested to USP Sterile Water for Injection Specifications, Corning™ Product Code.: 15373581

Cell Culture Grade Water Tested to USP Sterile Water for Injection Specifications, Corning™
BIOPHARMA

Product Code. 15373581
6 x 1 L, Bottle
Click to view available options
Quantity:
6 x 100 mL
6 x 500 mL
20 L
6 x 1 L
1 x 100 L
1 x 200 L
Packaging:
Bioprocess Container
Bottle
This item is not returnable. View return policy
Manufactured under cGMP (21 CFR 820) in ISO 13485 certified facility

Product Code. 15373581

Brand: Corning Media Tech 25055CM

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Manufactured under cGMP (21 CFR 820) in ISO 13485 certified facility

High quality water for use in cell culture applications. Corning™ Cell Culture Grade Water Tested to USP Sterile Water are used in preparation of purification and chromatography buffers.

Cell culture grade water is high-quality water suitable for use as a solvent in the preparation of cell culture media and other reagents for sensitive applications. Our bioprocess container selections are suitable for large-scale applications such as cleaning and upstream and downstream reagent preparation. Cell culture grade water goes through a wide array of testing which includes testing to the chemical and physical properties found in the USP and EP monographs for sterile Water for Injection (WFI).

Order Info

Supplied in a plastic bottle except MT-25-055-LB and -LH, which come in flexible media bags.

TRUSTED_SUSTAINABILITY

Specifications

Packaging Bottle
Quantity 6 x 1 L

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number33024005 Certificate TypeCertificate of Quality Date03/04/2025 Catalog Number25-055-CM
Lot Number26824014 Certificate TypeCertificate of Quality Date30/01/2025 Catalog Number25-055-CM
Lot Number19724006 Certificate TypeCertificate of Quality Date26/12/2024 Catalog Number25-055-CM
Lot Number19124014 Certificate TypeCertificate of Quality Date07/11/2024 Catalog Number25-055-CM
Lot Number24324002 Certificate TypeCertificate of Analysis Date10/03/2025 Catalog Number25-055-CM
  • Corning Matrigel , Corning Biocoat , Corning CoolProduct , Corning Media Tech , Corning
    Corning Lab Products Offer
    Promo Code24073

    Expires2025-04-25

  • Multiple Manufacturers
    Explore our new promotion and get up to 70% off some of our most popular products.
    Promo Code: 24127

    Expires2025-05-31

    Days

    43

    Hours

    10

    Minutes

    43

    Seconds

    50
  • Multiple Manufacturers
    Use promotion code 24138 to get amazing savings on products for your analysis
    Promo Code: 24138

    Expires2025-04-30

    Days

    12

    Hours

    10

    Minutes

    43

    Seconds

    50
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cell Culture Grade Water Tested to USP Sterile Water for Injection Specifications, Corning™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.