Learn More
Corning™ DeckWorks™ Bulk Pipet Tips
Compatible with DeckWorks racks and packaged in resealable polyethylene bags
Brand: Corning™ 4114
This item is not returnable.
View return policy
Description
Corning DeckWorks bulk pipet tips are compatible with DeckWorks racks and packaged in resalable polyethylene bags. Tip sizes include 10 μL, 200 μL, and 1000 μL and offer maximum savings.
Specifications
100 to 2000 μL | |
Natural | |
Round | |
DeckWorks™ Racks | |
DeckWorks |
Non-sterile | |
Help reduce the production and waste of plastic materials | |
Single-use | |
Yes | |
Polypropylene |
Product Suggestions
Customers who viewed this item also viewed.
Product Content Correction
Product Title
Corning™ DeckWorks™ Bulk Pipet Tips > Natural; 100-1000 μL
Spot an opportunity for improvement?Share a Content Correction