Learn More
Corning™ DeckWorks™ Bulk Pipet Tips
Compatible with DeckWorks racks and packaged in resealable polyethylene bags
Brand:  Corning™ 4114
24.24 GBP valid until 2025-04-30
Use promo code "24138" to get your promotional price.
 View more versions of this product
Product Code. 11931841
  • £30.50 / Pack of 768
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Corning DeckWorks bulk pipet tips are compatible with DeckWorks racks and packaged in resalable polyethylene bags. Tip sizes include 10 μL, 200 μL, and 1000 μL and offer ­maximum savings.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
100 to 2000 μL
Natural
Round
DeckWorks™ Racks
DeckWorks
Non-sterile
Help reduce the production and waste of plastic materials
Single-use
Yes
Polypropylene
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
  • Multiple Manufacturers
    Use promotion code 24138 to get amazing savings on products for your analysis
    Promo Code: 24138
    Expires2025-04-30
    Days
    5
    Hours
    14
    Minutes
    8
    Seconds
    15
Product Content Correction
Product Title
Corning™ DeckWorks™ Bulk Pipet Tips > Natural; 100-1000 μL