Learn More
Corning™ Polypropylene Plates
Manufactured from polypropylene for optimum visibility and high chemical resistance. Corning™ Polypropylene Plates are an excellent choice for transport and long-term storage.
Brand: Corning™ 3656
382.85 GBP valid until 2025-07-11
Use promo code "25721" to get your promotional price.
Description
Corning polypropylene microplates offer both small volume and large volume (blocks) well formats to meet assay and storage requirements.
- Flat, round, or V-shaped well bottom
- Features uniform skirt heights for greater robotic gripping surface
- Solvent resistant polypropylene provides compatibility with many common organic solvents (e.g., DMSO, ethanol, methanol)
- RNase-/DNase-free
- Available sterile or nonsterile
Order Info
Seal with Storage Mat III (sold separately).

Specifications
Standard | |
16 x 24 | |
Polypropylene | |
Sterile | |
Round | |
100/Cs. | |
100/Cs. |
Without Lid | |
Clear | |
384 | |
Non-treated | |
Transport and longterm storage | |
25/Pk. |
Special Offers
-
Corning Matrigel
,
Corning Biocoat
,
Corning CoolProduct
,
Corning Media Tech
,
Corning
-
Multiple Manufacturers
Spotlight
Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.Promo Code: 24155Expires: 2025-04-30
-
Multiple Manufacturers
Back to the Lab Summer Semester
Save up to 94% on Teaching Lab EssentialsPromo Code: 25721Expires: 2025-05-01
New Offer Save Now
Your input is important to us. Please complete this form to provide feedback related to the content on this product.