Learn More
Thermo Scientific™ CryoMed™ Controlled-Rate Freezer, General Purpose
Thermo Scientific CryoMed Controlled-Rate Freezers (CRF) are designed with reliability, protection, and data traceability in mind for your sample preparation needs.
Brand: Thermo Scientific™ TSCM17XV
21749.46 GBP valid until 2025-03-31
Use promo code "24119" to get your promotional price.
Description
For customers in research, cell and gene therapy, vaccine production, and biobanking, the CryoMed Controlled-Rate Freezer (CRF) provides precise, repeatable freezing results that protect samples from intracellular freezing. The CryoMed CRF features enhanced data traceability via a touch screen user interface and offers customizable freezing profiles while supporting 21 CFR part 11 and GMP needs.
Thermo Scientific CryoMed Controlled-Rate Freezers (CRF) are designed with reliability, protection, and data traceability in mind for your sample preparation and GMP production needs.
User-friendly operation with enhanced data traceability
- Intuitive touchscreen display allows for easy set-up, operation, and review of a freezing run
- Six pre-set freezing profiles and space for up to 14 user-defined, “custom” freeze profiles
- Integrated touchscreen UI logs usage and events to support 21 CFR part 11 requirements
- USB data export of .pdf file run logs support 21 CFR part 11
- User security: 3 levels of user accessibility
- “Run last” feature allows the same profile to be run consecutively with the touch of a button
- PC interface software provides remote operation capabilities, run review, and custom profile creation
Real-time run monitoring for sample protection
- Chamber and sample temperatures are monitored by Type T thermocouples, eliminating lag time and providing “real time” responsiveness
- Standard alarms to alert users of thermocouple failures, heater malfunction, high/low temperature limits, temperature tracking, power failure, and completion of run
- Remote alarm contacts
- Optional thermal printer
Reliable temperature performance for high throughput usage
- Dual solenoid valves are designed to balance LN2 injection volume for precise temperature control and accelerated freezing
- Preventative maintenance indicator for LN2 solenoid replacement limits downtime
- Temperature control and uniformity achieved via an air-handling and liquid nitrogen injection ring system
- Compatible with VHP cleaning techniques
- Environmentally friendly insulating foam
- Type 304 stainless steel with exterior powder-coat finish
Warranty varies by region. Check with your local representative for more information

Specifications
Controlled-rate freezer | |
17 L | |
Stainless Steel | |
CE | |
Warranty varies by region. Contact your local sales representative for more information | |
Front-loading | |
13 x 7 x 12 in. (33 x 17.8 x 30.5 cm) | |
Yes | |
Thermo Fisher Connect compatible | |
EU | |
<380 | |
69.8 kg |
0.6 cu. ft. | |
-180°C to +50°C | |
Powder-coated Steel | |
Touchscreen User Interface | |
General Purpose without Printer | |
24.3 x 37.3 x 21.7 in. (61 x 94.7 x 55.1 cm) | |
USB data export | |
PC Interface Software Supported | |
5.5A | |
220-230 V, 50/60Hz | |
154 lb. |
Product Suggestions
Customers who viewed this item also viewed
Special Offers
- Multiple Manufacturers
Your input is important to us. Please complete this form to provide feedback related to the content on this product.