missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cryopreserved Rat Kupffer Cells
Cryopreserved Rat Kupffer Cells
Brand: Gibco RTKCCS
This item is not returnable.
View return policy
Description
These cryopreserved purified Kupffer cells are isolated from adult male Sprague-Dawley rats. Each vial contains a minimum of one million cells.Easy to use cryopreserved Kupffer cells provide a convenient way to produce custom cell ratios for hepatic co-culture.
Each lot of Cryopreserved Rat Kupffer Cells is tested for:
Kupffer Cells for Improved Physiological Modeling
Growing evidence shows that under both normal and pathological conditions, many hepatocyte functions are regulated by substances released from neighboring non-parenchymal cells (NPC). These cells, particularly Kupffer cells, play an important role in the modulation of xenobiotic metabolism in the liver. Kupffer cells secrete potent mediators of the inflammatory response that controls liver inflammation. These cytokine mediators control hepatocyte metabolic rates through direct interactions with phase I and phase II enzymes. Kupffer cell and hepatocyte co-cultures can self-assemble within 72 hours of treatment with pro-inflammatory cytokines or LPS, and can function by effectively modulating P450 expression, thus giving a more physiologically relevant result. Studies indicate that co-culture of hepatocytes with NPCs would better represent both normal liver physiology as well as disease states.
For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.,
Note:
These products require shipment in LN2 vapor dewars therefore additional fees may apply.

Specifications
Passage 0 | |
Macrophage Cells | |
Cryopreserved | |
Rat | |
Normal | |
Tube | |
CellzDirect™ | |
1 vial |
1 vial of 1 million viable rat (Sprague-Dawley) Kupffer cells. Store in a cryostorage dewar or −135°C freezer. |
|
Adherent Cell Culture | |
Male | |
Liver | |
Pooled | |
1 x 106 | |
Rat Kupffer Cell | |
Purity, Response to Activation with LPS, Adherence, Minimum 1 Million Cell per Vial |
Product Sheet: Cryopreserved Human and Rat Kupffer Cells
Version: 2.0
File size: 147.2 kB
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cryopreserved Rat Kupffer Cells > 1 vial
Spot an opportunity for improvement?Share a Content Correction