Learn More
Asecos™ CS-CLASSIC Two-Door Yellow Safety Storage Cabinet
For the safe and proper storage of non-flammable chemicals and toxic substances in working areas.
Brand:  Asecos™ 3049504030508
 View more versions of this product
Product Code. 15896024
  • £941.00 / Each
Explore more special offers
Description
Description
  • Robust construction and longevity – sturdy all-welded construction for high industrial requirements
  • High-quality, scratch-resistant outer body
  • Double-walled door construction
  • In accordance with TRGS 510
  • Available with double or single wing doors
  • Modern bicolor design
  • Ventilation – ready for connection to a technical exhaust system (DN 75), natural ventilation at the cabinet bottom
  • Adjusting aids are integrated in the cabinet base, fast and effective compensation for uneven floors
  • Interior equipment available includes height-adjustable shelves, pull-out shelves and bottom collecting sumps
  • Powder-coated textured surface
  • No unauthorized use – doors lockable with cylinder locking, integrated into the turning handle
Specifications
Specifications
TRGS 510
493 x 996 x 1835 mm
Wing
3 x Shelves: 100 kg each, Insert: 60 kg, Sump: 33 L, Maximum load: 600 kg
Powder-coated
Yellow
Safety storage cabinet
3 x 100 kg galvanized sheet steel shelves; 1 x 60 kg galvanized sheet steel insert; 1 x 33 L galvanized sheet steel sump
520 x 1055 x 1950 mm
Storage of non-flammable and toxic substances
2
110 kg
Steel
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Asecos™ CS-CLASSIC Two-Door Yellow Safety Storage Cabinet > 3x 100 kg shelves; 1x 60 kg insert; 1x 33 L sump