missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CSPD™ Substrate (0.25 mM Ready-To-Use)

This CSPD™ substrate is a chemiluminescent alkaline phosphatase substrate, ready-to-use for protein or nucleic acid blotting on nitrocellulose membranes.

Brand:  Invitrogen™ T2142

 View more versions of this product

Product Code. 12644325

  • £236.00 / Each
Estimated Shipment: 31-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

This CSPD™ substrate is a chemiluminescent alkaline phosphatase substrate, ready-to-use for protein or nucleic acid blotting on nitrocellulose membranes. This substrate can also be used in solution-based assays.

This CSPD™ chemiluminescent substrate for alkaline phosphatase let you detect alkaline phosphatase and alkaline phosphatase-labeled molecules with unparalleled sensitivity, speed, and ease.

  • Chemiluminescent substrates provide highly sensitive replacements for the widely used fluorogenic substrate methylumbelliferyl phosphate (MUP), and the colorimetric substrate p-nitrophenyl phosphate (pNPP).
  • Low background luminescence coupled with high-intensity light output enables detection of alkaline phosphatase labels with the highest possible sensitivity and signal-to-noise.

This versatile chemiluminescent substrate exhibits high sensitivity in membrane-based applications such as Southern, Northern, and Western blotting. This substrate can also be used in solution-based assays such as immunoassays, DNA probe assays, enzyme assays, and reporter gene assays. Maximum light levels are reached in approximately 10 minutes and glow emission persists for several hours.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

ELISA Substrate, CSPD™ Substrate
Chemiluminescence
Liquid
Bottle, Bottles
CSPD™, NovaBright™
Store the product at 4°C.
4°C
100 mL, 100mL
100mL
Protein assays and analysis, southern, northern, western blotting, can also be used in solution-based assays such as immunoassays, DNA probe assays, enzyme assays, and reporter gene assays, protein biology, cell analysis, enzyme reporter assays, ELISA, alkaline phosphatase assays, build your own immunoassay, ready-to-use immunoassay
AP (Alkaline Phosphatase) Substrate, Alkaline Phosphatase
CSPD™ Substrate, 100mL, Liquid Form, Chemiluminescent Alkaline Phosphatase Substrate, 0.25mM Ready-to-use For Protein or Nucleic Acid Blotting on Nitrocellulose membranes, Protein Assays and Analysis, Southern, Northern and Western blotting, Can also
Room Temperature
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2411071 Certificate TypeCertificate of Analysis Date13/01/2025 Catalog NumberT2142
Lot Number2106064 Certificate TypeCertificate of Analysis Date15/07/2021 Catalog NumberT2142
Lot Number2103063 Certificate TypeCertificate of Analysis Date29/03/2021 Catalog NumberT2142
Lot Number2011062 Certificate TypeCertificate of Analysis Date08/12/2020 Catalog NumberT2142
Lot Number2008061 Certificate TypeCertificate of Analysis Date28/08/2020 Catalog NumberT2142
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ CSPD™ Substrate (0.25 mM Ready-To-Use) > 100 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.