missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ambion™ Custom Select siRNA
Custom Select siRNA
Brand: Ambion™ 4390828
This item is not returnable.
View return policy
Description
Silencer™ Select custom siRNAs (LNA-modified 21-bp duplex with overhangs) are RNA interference (RNAi) tools ideal for knocking down gene expression in order to study protein function in a wide range of cell types. These siRNAs provide high knock-down efficiencies and specificities.
Specifications
Contains dried siRNA and Nuclease-free Water (1.75 mL), shipped at ambient temperature. Upon receipt, store siRNAs in a non-frost-free freezer at or below –20 °C | |
Knocking down gene expression in order to study protein function in a wide range of cell types | |
Custom Select siRNA | |
20 nmol |
Custom Select siRNA, 20nmol, Standard Purity, ≥80% Purity, Ideal for Knocking Down Gene Expression in Order to Study Protein Function in a Wide Range of Cell Types, These siRNAs Provide High Knock-down Efficiencies and Specificities | |
Ambion™, Silencer™ Select | |
Standard |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Protocol: Silencer® Select Pre-designed, Validated, and Custom siRNAs
Version: 1.0
File size: 1.7 MB
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ambion™ Custom Select siRNA > 20 nmol
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction