missing translation for 'onlineSavingsMsg'
Learn More
Cyclohexene oxide, 98+%, Thermo Scientific Chemicals Product Code.: 11464584

Cyclohexene oxide, 98+%, Thermo Scientific Chemicals

Product Code. 11464584
500 g
Click to view available options
Quantity:
100 g
500 g
2500 g
This item is not returnable. View return policy

Product Code. 11464584

Brand: Thermo Scientific Alfa Aesar A13185.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It is an important raw material and intermediate used in organic synthesis, dyestuffs and in agrochemical industries. It is used in medicine. Cyclohexene oxide (epoxycyclohexane) is an useful monomer in polymerization and coating industry. It is used in the synthesis of alicyclic target materials including pharmaceuticals, perfumery and dyestuffs. It is used as a monomer in photopolymerizations, with carbonmonoxide to yield aromatic polycarbonates which has minimum impurities.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, dyestuffs and in agrochemical industries. It is used in medicine. Cyclohexene oxide (epoxycyclohexane) is an useful monomer in polymerization and coating industry. It is used in the synthesis of alicyclic target materials including pharmaceuticals, perfumery and dyestuffs. It is used as a monomer in photopolymerizations, with carbonmonoxide to yield aromatic polycarbonates which has minimum impurities.

Solubility
Insoluble in water

Notes
Store at room temperature. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 286-20-4
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.145
MDL Number MFCD00005162
InChI Key ZWAJLVLEBYIOTI-UHFFFAOYSA-N
Synonym cyclohexene oxide, 7-oxabicyclo 4.1.0 heptane, 1,2-epoxycyclohexane, cyclohexene epoxide, cyclohexylene oxide, tetramethyleneoxirane, cyclohexene 1-oxide, epoxycyclohexane, cyclohexeneoxide, 1,2-cyclohexene oxideShow MoreShow Less
PubChem CID 9246
IUPAC Name 7-oxabicyclo[4.1.0]heptane
SMILES C1CCC2C(C1)O2

Specifications

Melting Point -40°C
Density 0.972
Boiling Point 129°C to 130°C
Flash Point 24°C (75°F)
Refractive Index 1.452
Quantity 500 g
UN Number UN2920
Beilstein 383568
Solubility Information Insoluble in water
Formula Weight 98.15
Percent Purity ≥98%
Chemical Name or Material Cyclohexene oxide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10252543 Certificate TypeCertificate of Analysis Date23/09/2024 Catalog NumberA13185.22,A13185.36,A13185.0E
Lot NumberA0451551 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberA13185.22,A13185.36,A13185.0E
Lot NumberA0457806 Certificate TypeCertificate of Analysis Date20/08/2024 Catalog NumberA13185.22,A13185.36,A13185.0E
Lot NumberA10098753 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA13185.22,A13185.36,A13185.0E
Lot Number10235116 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA13185.22,A13185.36,A13185.0E

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cyclohexene oxide, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.