missing translation for 'onlineSavingsMsg'
Learn More
Cyclohexyl hexanoate, 99%, Thermo Scientific Chemicals Product Code.: 11447124

Cyclohexyl hexanoate, 99%, Thermo Scientific Chemicals

Product Code. 11447124
100 g
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy

Product Code. 11447124

Brand: Thermo Scientific Alfa Aesar B21547.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6243-10-3
MDL Number MFCD00046356

Specifications

Molecular Formula C4H9NO
Refractive Index 1.446
Quantity 100 g
Synonym 3-oxetanemethanamine, 3-aminomethyl-oxetane, 3-aminomethyloxetane, 1-oxetan-3-yl methanamine, c-oxetan-3-yl-methylamine, oxetan-3-ylmethamine, oxetane-3-methanamine, n-methyl-3-oxetanamne, oxetane-3-ylmethanamine, pubchem23208
InChI Key KTHZBRAXOLUNBN-UHFFFAOYSA-N
SMILES C1C(CO1)CN
IUPAC Name oxetan-3-ylmethanamine
Molecular Weight (g/mol) 87.12
PubChem CID 46840007
Formula Weight 198.31
Percent Purity 99%
Chemical Name or Material Cyclohexyl hexanoate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberE3746A Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB21547.14,B21547.22
Lot Number10207541 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB21547.14,B21547.22
Lot Number10216181 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB21547.14,B21547.22
Lot Number10211764 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB21547.14,B21547.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cyclohexyl hexanoate, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.