Learn More
Invitrogen™ CytoTune™ EmGFP Sendai Fluorescence Reporter
The CytoTune™ EmGFP Sendai Fluorescence Reporter helps you determine whether the Sendai particles that are used in all CytoTune™-iPS Sendai reprogramming kits can transduce into a given cell type before you advance to one of these kits.
Brand: Invitrogen™ A16519
Description
This vector expresses the next-generation Aequorea victoria EGFP gene, encoding the emerald green fluorescent protein (EmGFP). The cell will express the EmGFP, which is bright, photostable, and ideal for testing transduction.
The CytoTune™ EmGFP Sendai Fluorescence Reporter enables a better understanding of how both the original CytoTune™-iPS Sendai Reprogramming kits and the newer CytoTune™-iPS 2.0 Sendai Reprogramming Kit transduces various cell types. The Sendai particle enters a cell via its HN envelope protein that recognizes sialic acid and is widely expressed in mammalian cells, allowing Sendai virus to target a wide range of cell types. The Sendai virus vectors do not enter the nucleus, thus avoiding any DNA phase, as the viral genome remains as RNA in the cytoplasm.

Specifications
Sendai Reporter | |
GFP (EmGFP) | |
Sendai virus | |
CytoTune™ | |
Liquid |
GFP (EmGFP) | |
1 Tube, store (at -68°C to -85°C.) | |
Proprietary | |
1 Pack |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
LG190074 | Certificate of Analysis | 12/02/2025 | A16519 |
LG200076 | Certificate of Analysis | 12/02/2025 | A16519 |
LG190072 | Certificate of Analysis | 11/04/2022 | A16519 |
LG180065 | Certificate of Analysis | 22/03/2021 | A16519 |
LG180062 | Certificate of Analysis | 18/01/2021 | A16519 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.