Learn More
Invitrogen™ Yeast tRNA
Supplied in lyophilized form
Brand: Invitrogen™ 15401029
Description
Yeast tRNA is suitable for use as a carrier in nucleic acid purification and precipitation procedures.
Array CGH, Gene Expression Analysis and Genotyping, Genotyping and Genomic Profiling, Microarray Analysis
Order Info
Shipping Condition: Approved for shipment at Room Temperature or on Wet or Dry Ice

Specifications
Yeast tRNA | |
Yeast tRNA | |
Carrier for Nucleic Acid Precipitation | |
Lyophilized Powder |
Store in freezer (-5 to -30°C). | |
Approved for shipment at Room Temperature or on Wet or Dry Ice | |
50 mg |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
3072400 | Certificate of Analysis | 31/01/2025 | 15401029 |
2601321 | Certificate of Analysis | 29/01/2025 | 15401029 |
2215247 | Certificate of Analysis | 29/01/2025 | 15401029 |
2525508 | Certificate of Analysis | 29/01/2025 | 15401029 |
2779690 | Certificate of Analysis | 29/01/2025 | 15401029 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.