missing translation for 'onlineSavingsMsg'
Learn More
D-(+)-Biotin, 98+%, Thermo Scientific Chemicals Product Code.: 11354529

D-(+)-Biotin, 98+%, Thermo Scientific Chemicals

Product Code. 11354529
10 g
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
This item is not returnable. View return policy

Product Code. 11354529

Brand: Thermo Scientific Alfa Aesar A14207.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Biotin is a form of vitamin B that helps the body to break down fats and carbohydrates. It is used as an alternative medicine to treat or prevent biotin deficiency, which is caused by malnutrition, rapid weight loss, long-term tube feeding and other medical conditions. It is an essential vitamin which is important for amino acid and energy metabolism.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in hot water, dimethyl sulfoxide, alcohol and benzene.

Notes
Store in cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58-85-5
Molecular Formula C10H16N2O3S
Molecular Weight (g/mol) 244.31
MDL Number MFCD00005541
InChI Key YBJHBAHKTGYVGT-UHFFFAOYNA-N
Synonym biotin, d-biotin, vitamin h, coenzyme r, vitamin b7, bios ii, factor s, bioepiderm, d +-biotin, biodermatin
PubChem CID 171548
ChEBI CHEBI:15956
IUPAC Name 5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]pentanoic acid
SMILES OC(=O)CCCCC1SCC2NC(=O)NC12

Specifications

Melting Point 224°C to 226°C
Assay Percent Range ≥98%
Quantity 10 g
Beilstein 86838
Sensitivity Light sensitive
Merck Index 14,1231
Solubility Information Soluble in hot water,dimethyl sulfoxide,alcohol and benzene.
Optical Rotation +91° (c=1 in 0.1N NaOH)
Formula Weight 244.32
Percent Purity ≥98%
Chemical Name or Material D-(+)-Biotin
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberN24L039 Certificate TypeCertificate of Analysis Date26/02/2025 Catalog NumberA14207.06,A14207.14,A14207.03,A14207.09
Lot Number10253363 Certificate TypeCertificate of Analysis Date11/12/2024 Catalog NumberA14207.06,A14207.03,A14207.14,A14207.09
Lot Number10252481 Certificate TypeCertificate of Analysis Date18/09/2024 Catalog NumberA14207.06,A14207.03,A14207.14,A14207.09
Lot Number10251062 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberA14207.06,A14207.03,A14207.14,A14207.09
Lot Number10252240 Certificate TypeCertificate of Analysis Date29/08/2024 Catalog NumberA14207.06,A14207.03,A14207.14,A14207.09

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
D-(+)-Biotin, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.