missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals D(-)-Ribose, 99+% Product Code.: 10320164

Thermo Scientific Chemicals D(-)-Ribose, 99+%

Product Code. 10320164
5 g, Glass bottle
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10320164

Brand: Thermo Scientific Chemicals 132360050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-69-1
Molecular Formula C5H10O5
Molecular Weight (g/mol) 150.13
MDL Number MFCD00135453
InChI Key HMFHBZSHGGEWLO-ZHZWXSSZNA-N
Synonym 2r,3r,4r-2,3,4,5-tetrahydroxypentanal, aldehydo-d-ribose, ribose, d, d--ribose, aldehydo-d-ribo-pentose, ribo-2,3,4,5-tetrahydroxyvaleraldehyde, d, d-ribo-2,3,4,5-tetrahydroxyvaleraldehyde, ribose 9ci, dsstox_cid_23917, dsstox_gsid_43917Show MoreShow Less
PubChem CID 5311110
ChEBI CHEBI:47014
SMILES OC[C@H]1OC(O)[C@H](O)[C@@H]1O

Specifications

Melting Point 88.0°C to 92.0°C
Color Beige to White or Yellow
Loss on Drying 1% max. (40°C, 5 hrs, 20 torr)
Infrared Spectrum Authentic
Assay Percent Range 99% min. (ELSD) (HPLC)
Packaging Glass bottle
Quantity 5 g
Beilstein 01, I, 434
Merck Index 15, 8328
Specific Rotation Condition − 20.00 (20.00°C c=4,H2O)
Specific Rotation − 20.00
IUPAC Name (2R,3R,4R)-2,3,4,5-tetrahydroxypentanal
Formula Weight 150.13
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material D(-)-Ribose
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0464130 Certificate TypeCertificate of Analysis Date09/07/2024 Catalog Number132361000,132360250,132365000,132360050
Lot NumberA0460565 Certificate TypeCertificate of Analysis Date28/02/2024 Catalog Number132361000,132360250,132365000,132360050
Lot NumberA0451350 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number132361000,132360250,132365000,132360050
Lot NumberA0407968 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number132361000,132360250,132365000,132360050
Lot NumberA0410792 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number132361000,132360250,132365000,132360050
Product Identifier
  • D(-)-Ribose

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals D(-)-Ribose, 99+% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.