missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals D(+)-Trehalose dihydrate, 99% Product Code.: 10483245

Thermo Scientific Chemicals D(+)-Trehalose dihydrate, 99%

Product Code. 10483245
100 g, Glass bottle
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic Bottle
This item is not returnable. View return policy

Product Code. 10483245

Brand: Thermo Scientific Chemicals 182551000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6138-23-4
Molecular Formula C12H22O11·2H2O
Molecular Weight (g/mol) 378.32
MDL Number MFCD00071594
InChI Key DPVHGFAJLZWDOC-PVXXTIHASA-N
Synonym trehalose dihydrate, d-+-trehalose dihydrate, a,a-trehalose, d +-trehalose dihydrate, unii-7yin7j07x4, alpha,alpha-trehalose dihydrate, d-trehalose dihydrate, mycose,1-o-alpha-d-glucopyranosyl-alpha-d-glucopyranoside, alp,alp.-trehalose, 2r,3s,4s,5r,6r-2-hydroxymethyl-6-2r,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl tetrahydropyran-2-yl oxy-tetrahydropyran-3,4,5-triolShow MoreShow Less
PubChem CID 181978
IUPAC Name (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyoxane-3,4,5-triol;dihydrate
SMILES C(C1C(C(C(C(O1)OC2C(C(C(C(O2)CO)O)O)O)O)O)O)O.O.O

Specifications

Melting Point 97.0°C to 99.0°C
Assay Percent Range 98.5% min. (ELSD) (HPLC)
Packaging Glass bottle
Quantity 100 g
Merck Index 159742
Specific Rotation Condition + 179.00 (20.00°C c=2,H2O)
Specific Rotation + 179.00
Solubility Information (10% in water) Clear colorless
Formula Weight 378.32
Percent Purity 99%
Chemical Name or Material D(+)-Trehalose dihydrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0466907 Certificate TypeCertificate of Analysis Date30/10/2024 Catalog Number182555000,182550025,182550250,182551000
Lot NumberA0465663 Certificate TypeCertificate of Analysis Date23/08/2024 Catalog Number182555000,182550025,182550250,182551000
Lot NumberA0463749 Certificate TypeCertificate of Analysis Date18/06/2024 Catalog Number182555000,182550025,182550250,182551000
Lot NumberA0461546 Certificate TypeCertificate of Analysis Date31/03/2024 Catalog Number182555000,182550025,182550250,182551000
Lot NumberA0460466 Certificate TypeCertificate of Analysis Date22/02/2024 Catalog Number182555000,182550025,182550250,182551000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals D(+)-Trehalose dihydrate, 99% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.