missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals D-(+)-Trehalose dihydrate, 99% Product Code.: 11404027

Thermo Scientific Chemicals D-(+)-Trehalose dihydrate, 99%

Product Code. 11404027
5 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11404027

Brand: Thermo Scientific Chemicals A19434.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

D-(+)-Trehalose dihydrate is used as a food additive in many processed foods and pharmaceutical excipients. It is widely used as a direct replacement of conventional sweeteners and protects the cells from various environmental stresses. It finds application in the treatment of Huntington's disease and Parkinson's disease.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water and hot alcohol.

Notes
Store in cool place. Hygroscopic. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6138-23-4
Molecular Formula C12H22O11
Molecular Weight (g/mol) 342.297
MDL Number MFCD00071594
InChI Key HDTRYLNUVZCQOY-LIZSDCNHSA-N
Synonym trehalose, d-trehalose, alpha,alpha-trehalose, mycose, ergot sugar, alpha-d-trehalose, d-+-trehalose, d-+-trehalose, anhydrous, alpha,alpha'-trehalose, trehaoseShow MoreShow Less
PubChem CID 7427
ChEBI CHEBI:16551
IUPAC Name (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyoxane-3,4,5-triol
SMILES C(C1C(C(C(C(O1)OC2C(C(C(C(O2)CO)O)O)O)O)O)O)O

Specifications

Melting Point 97°C to 99°C
Assay Percent Range 0.99
Quantity 5 g
Beilstein 5322018
Sensitivity Hygroscopic
Solubility Information Soluble in water and hot alcohol.
Formula Weight 378.33 (342.29 Anhydrous)
Chemical Name or Material D-(+)-Trehalose dihydrate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10250047 Certificate TypeCertificate of Analysis Date05/04/2024 Catalog NumberA19434.06,A19434.14,A19434.22
Lot Number10247011 Certificate TypeCertificate of Analysis Date16/06/2023 Catalog NumberA19434,A19434.06,A19434.14,A19434.22
Lot Number10242382 Certificate TypeCertificate of Analysis Date10/08/2022 Catalog NumberA19434.06,A19434.14,A19434.22
Lot Number10189935 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA19434.06,A19434.14,A19434.22
Lot Number10157886 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA19434.06,A19434.14,A19434.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals D-(+)-Trehalose dihydrate, 99% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.