Learn More
Thermo Scientific™ Deep Vacuum Oil Pumps for Vacuum Concentrators
Achieve maximum solute recovery with the Thermo Scientific™Deep Vacuum Oil Pump.
Brand:  Thermo Scientific™ VLP120-230
 View more versions of this product
Product Code. 13162053
  • £3960.00 / Each
Explore more special offers
Description
Description
Achieve maximum solute recovery with the Thermo Scientific™ Deep Vacuum Oil Pumps for Vacuum Concentrators, designed to work with all Thermo Scientific™ SpeedVac™ concentrating and freeze drying systems.
Features:
  • Three-position gas ballast control optimizes pump performance to suit all applications
  • Large, sideview sight glass allows for convenient checking of proper oil levels at a glance
All Rotary Vane Oil Pumps come standard with:
  • Muffler (OME190 Oil Mist Eliminator)
  • Two Oil Filters (MF190)
  • Quick Drain Kit
TRUSTED_SUSTAINABILITY
Specifications
Specifications
VLP120; 230V 50Hz
116L/min. (at 60Hz); 97L/min. (at 50Hz)
1.5 mtorr/1.95 x10³ mbar
6 in.
9 in.
17 in.
48 lb.
230 V 50 Hz
50 Hz
Deep Vacuum Oil Pump
0.7 L
12.7 OD
160 mm
230 mm
420 mm
22 kg
230 V
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Specification Sheets
Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Deep Vacuum Oil Pumps for Vacuum Concentrators >