missing translation for 'onlineSavingsMsg'
Learn More
Honeywell Safety Products™ Dexgrip Carded Gloves Product Code.: 12665193
In Stock
to see stock.
Add to basket

Honeywell Safety Products™ Dexgrip Carded Gloves

Product Code. 12665193
8
Click to view available options
Size:
7
8
9

Product Code. 12665193

Brand: Honeywell Safety Products™ 209414008

In Stock
to see stock.
Add to basket
Add to basket

Comfortable, hand conforming and snug, this knit glove provides both breathability and grip for handling of damp or non-greasy parts. Honeywell Safety Products™ Dexgrip Carded Gloves come in effective, single-pair packaging designed for a self-service environment.

  • Knitted cotton/polyamide grey glove.
  • Palm and fingertips coated in blue, crinkled latex for abrasion and perforation resistance as well as excellent grip.
  • Conforms snugly and provides good dexterity.
  • Cotton thread absorbs perspiration.
  • Ventilated back provides breathability.
  • Product emphasized in effective, single-pair packaging designed for a self-service environment.
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE CATEGORY II Intermediary risks, EN 420-2003 Basic standard, EN 388-2003 Mechanical risks, EN 407-2004 Thermal risks, EC Category PPE 2, Quality Assurance ISO 9001 / 2000, EC Certificate Number 0075/797/162/12/08/0519, EC Attestation Number 0075/797/162/12/08/0519
Classification Dexterity: 5
Coated Area Palm, Fingertips
Description Gauge 10
Coating Material Crinkled Latex
Color Gray, Blue
Cut Resistance Rating 1
Material Polyamide, Latex, Cotton
Product Type Gloves
Size 8
Thickness (Metric) 1.5 to 1.8 mm
For Use With (Application) Agriculture, Building, Construction, Glass, Green Spaces, Logistics, Transportation and Wood industries
Packaging Type Single pair
Quantity 10Pair/PK
Tear Rating 4
Length (Metric) 26 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Honeywell Safety Products™ Dexgrip Carded Gloves >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.