missing translation for 'onlineSavingsMsg'
Learn More
Diethyl sulfate, 98%, Thermo Scientific Chemicals Product Code.: 11309095

Diethyl sulfate, 98%, Thermo Scientific Chemicals

Product Code. 11309095
100 mL
Click to view available options
Quantity:
100 mL
500 mL
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

Product Code. 11309095

Brand: Thermo Scientific Alfa Aesar L14291.AE

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-67-5
Molecular Formula C4H10O4S
Molecular Weight (g/mol) 154.18
MDL Number MFCD00009099
InChI Key DENRZWYUOJLTMF-UHFFFAOYSA-N
Synonym sulfuric acid, diethyl ester, diethyl sulphate, diethylsulfate, diaethylsulfat, sulfuric acid diethyl ester, diethyl tetraoxosulfate, diaethylsulfat german, unii-k0fo4vfa7i, des van, diethylester kyseliny siroveShow MoreShow Less
PubChem CID 6163
ChEBI CHEBI:34699
IUPAC Name diethyl sulfate
SMILES CCOS(=O)(=O)OCC

Specifications

Melting Point -25°C
Density 1.177
Boiling Point 208°C (decomposition)
Flash Point 104°C (219°F)
Refractive Index 1.4005
Quantity 100 mL
UN Number UN1594
Beilstein 1209714
Merck Index 14,3130
Formula Weight 154.19
Percent Purity 98%
Chemical Name or Material Diethyl sulfate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10236400 Certificate TypeCertificate of Analysis Date08/03/2024 Catalog NumberL14291.AP,L14291.AE
Lot NumberA0443420 Certificate TypeCertificate of Analysis Date25/07/2023 Catalog NumberL14291.AP,L14291.AE
Lot NumberD26Z075 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL14291.AP,L14291.AE
Lot NumberS12E052 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL14291.AP,L14291.AE
Lot Number10179103 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL14291.AP,L14291.AE

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Diethyl sulfate, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.