missing translation for 'onlineSavingsMsg'
Learn More
Diethyldithiocarbamic acid, silver salt, 99%, for analysis, Thermo Scientific Chemicals Product Code.: 10112430

Diethyldithiocarbamic acid, silver salt, 99%, for analysis, Thermo Scientific Chemicals

Product Code. 10112430
25 g, Glass bottle
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10112430

Brand: Thermo Scientific Acros 114290250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1470-61-7
Molecular Formula C5H10AgNS2
Molecular Weight (g/mol) 256.13
MDL Number MFCD00004929
InChI Key NSVHDIYWJVLAGH-UHFFFAOYSA-M
Synonym silver diethyldithiocarbamate, unii-gvn001b86o, agddtc, silver n,n-diethylcarbamodithioate, diethyldithiocarbamic acid, silver salt, diethyl dithio carbamic acid silver salt, silver i diethyldithiocarbamate, silver, diethylcarbamodithioato-ks,ks', ditiocarb silver, silver n,n-diethyldithiocarbamateShow MoreShow Less
PubChem CID 3034078
IUPAC Name silver;N,N-diethylcarbamodithioate
SMILES CCN(CC)C(=S)[S-].[Ag+]

Specifications

Melting Point 172.0°C to 175.0°C
Color Green to Khaki
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula (C2H5)2NCS2Ag
Quantity 25 g
Beilstein 04, 121
Solubility Information (0.5% in pyridine) clear
Formula Weight 256.13
Percent Purity 99%
Grade Analysis
Physical Form Amorphous Powder
Chemical Name or Material Diethyldithiocarbamic acid, silver salt, For analysis
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0465524 Certificate TypeCertificate of Analysis Date05/09/2024 Catalog Number114290050,114291000,114290250
Lot NumberA0458772 Certificate TypeCertificate of Analysis Date21/02/2024 Catalog Number114290050,114291000,114290250
Lot NumberA0433368 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number114290050,114291000,114290250
Lot NumberA0398465 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number114290050,114291000,114290250
Lot NumberA0421382 Certificate TypeCertificate of Analysis Date29/07/2021 Catalog Number114290050,114291000,114290250
compliance-icons
Product Identifier
  • Diethyldithiocarbamic acid, silver salt
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Diethyldithiocarbamic acid, silver salt, 99%, for analysis, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.