Learn More
Cole-Parmer™ Digi-Sense™ USB Temperature and Temperature/RH Dataloggers
Digi-Sense™ USB Dataloggers are portable or mountable to fit multiple applications
Brand:  Cole-Parmer™ 20250-44
 View more versions of this product
Product Code. 15890006
  • £114.00 / Each
Explore more special offers
Description
Description
  • Each Digi-Sense data logger comes user ready with software, mounting clip and easy to following instructions.
  • Intended applications include monitoring pharmaceutical, food & beverage, storage and office, museum, laboratory and transportation environments.
  • Ease of use and affordability make these instruments ideal for any need where accurate temperature monitoring is needed.
  • Intuitive software supports two modes of data display and powerful data scaling and panning functionality. Data can be downloaded in spreadsheet compatible formats for analysis or graphically printed out.
  • The durable polycarbonate housing and precision sensors and electronics are backed by a 2-year warranty.
Services
2 Years
Specifications
Specifications
Datalogger
32,000 Readings
High/Low, Selectable
3.6 V Lithium, 1/2 AA
Software, mounting clip and hardware, protective plastic cover, and battery.
USB
Monitoring pharmaceutical, food & beverage, storage and office, museum, laboratory and transportation environments. 
±2°F (1°C)
0.01°
3.875 in.
0.4 kg
Status Indicator Lights
Not Calibrated
Setup, real-time monitoring, data acquisition, data analysis
1 Year Typical
1
Plastic
Type-K Thermocouple Probes
No humidity monitoring
-200°C to +1370°C
9.8 cm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Cole-Parmer™ Digi-Sense™ USB Temperature and Temperature/RH Dataloggers > USB Temperature Type K Datalogger; Display type: Status indicator lights