Learn More
Thermo Scientific™ DiluFlux™ Pro Automated Gravimetric Dilutor
Accurately dispense media into suitable containers according to a set dilution using this gravimetric diluter which allows the use of two diluents.
Brand: Thermo Scientific™ DB4150A
7588.32 GBP valid until 2025-04-13
Use promo code "24122" to get your promotional price.
Description
Accurately dispense broth medium into suitable containers for food testing using the Thermo Scientific™ DiluFlux™ Pro Automated Gravimetric Dilutor. DiluFlux Pro Dilutors are compact, easy-to-use, and designed to deliver precise quantities of diluents rapidly with the touch of a button, so that you can work more confidently and accurately. When combined with the Thermo Scientific™ Peristaltic Pump and Thermo Scientific™ Homogenizer, your lab bench will be equipped with a flexible automated culture media solution to increase your productivity.
Automate the dilution step of your Thermo Scientific™ Dry-Bags™ Enrichment Media, as well as other formats, for maximum efficiency. The easy-to-use DiluFlux Pro Gravimetric Dilutors allow the simultaneous use of two diluents by having dual dispensing heads that are fed by dual peristaltic pumps mounted into the unit.
- Intuitive: simply press two keys, tare and go, and the DiluFlux Pro Dilutor will automatically dilute your sample to the desired dilution factor
- Accurate: featuring Watson-Marlow™ peristaltic pumps, high-tech flow regulators, and top of the line weighing platforms for trusted reproducibility and repeatability
- Versatile: compatible with many prepared media formats, the easy-to clean, stainless steel DiluFlux Pro Dilutor is an ideal fit for any food testing laboratory
- High-quality: the DiluFlux Pro Dilutors are ISO 7218, ISO 6887-1, FDA BAM-compliant and are made in France under ISO 9001:2008 certification
Not all products are available for sale in all territories. Please inquire.

Specifications
DiluFlux Pro-5 Automated Gravimetric Dilutor w/ robotic dispensing arm | |
Automated Dilutor Dual Pump unit only | |
41.5 x 31 x 37.5 cm | |
31 cm | |
12.5 kg | |
100/240 V |
Dry-Bags | |
Stainless Steel (Case) | |
41.5 cm | |
37.5 cm | |
100/127 V 220/240 V 50/60 Hz | |
50/60 Hz |
Special Offers
-
Multiple Manufacturers
Precision at Your Fingertips
Master liquid handling with ease.Promo Code: 24122Expires: 2025-04-13
Days
11Hours
15Minutes
49Seconds
34 - Multiple Manufacturers
Your input is important to us. Please complete this form to provide feedback related to the content on this product.