Learn More
Thermo Scientific™ DNAPac™ PA200 Oligonucleotide HPLC Columns
BIOPHARMA
Enjoy strong anion exchange for high-resolution analysis and purification of synthetic oligonucleotides with Thermo Scientific™ DNAPac™ PA200 Oligonucleotide HPLC Columns.
£587.93 - £6612.90
Specifications
Pore Size Nonporous
Quantity Each
Temperature 85°C
Stationary Phase Quaternary Ammonium
Product Line DNAPac
View More Specs
Products 11
Product Code Brand Length (Metric) Diameter (Metric) Particle Size Price Quantity & Availability  
Product Code Brand Length (Metric) Diameter (Metric) Particle Size Price Quantity & Availability  
11341813
BIOPHARMA
View Documents Promotion Details
Thermo Scientific™
063000
250 mm 4 mm 8 μm
£1231.83
Each
In Stock
to see stock.
Edge
Add to basket
11361843
View Documents
Thermo Scientific™
063421
250 mm 9 mm 8 μm
£2582.85
Each
Estimated Shipment: 04-07-2025
to see stock.
Add to basket
11381843
View Documents
Thermo Scientific™
063425
250 mm 2 mm 8 μm
£1206.00
Each
Estimated Shipment: 07-05-2025
to see stock.
Add to basket
11331813
View Documents
Thermo Scientific™
062998
50 mm 4 mm 8 μm
£587.93
Each
Estimated Shipment: 08-05-2025
to see stock.
Add to basket
11351843
View Documents
Thermo Scientific™
063419
50 mm 9 mm 8 μm
£874.35
Each
Estimated Shipment: 08-05-2025
to see stock.
Add to basket
11371843
View Documents
Thermo Scientific™
063423
50 mm 2 mm 8 μm
£587.93
Each
Estimated Shipment: 18-07-2025
to see stock.
Add to basket
15139673
View Documents
Thermo Scientific™
082510
250 mm 4.6 mm 4 μm
£2376.83
Each
Estimated Shipment: 04-07-2025
to see stock.
Add to basket
15159673
BIOPHARMA
View Documents Promotion Details
Thermo Scientific™
082509
150 mm 4.6 mm 4 μm
£2288.99
Each
Estimated Shipment: 09-05-2025
to see stock.
Add to basket
15149673
View Documents
Thermo Scientific™
082508
50 mm 4.6 mm 4 μm
£1783.88
Each
Estimated Shipment: 15-05-2025
to see stock.
Add to basket
15510244
View Documents
Thermo Scientific™
088781
250 mm 22 mm 8 μm
£6612.90
Each
Estimated Shipment: 04-07-2025
to see stock.
Add to basket
15500244
View Documents
Thermo Scientific™
088780
50 mm 22 mm 8 μm
£2427.08
Each
Estimated Shipment: 14-05-2025
to see stock.
Add to basket
Description
Description
Enjoy strong anion exchange for high-resolution analysis and purification of synthetic oligonucleotides with Thermo Scientific™ DNAPac™ PA200 Oligonucleotide HPLC Columns.
  • Achieve n, n-1 resolution for oligonucleotides
  • Resolve oligonucleotides with secondary structures
  • Assay phosphorothioate purity
  • Selectivity control with eluent pH, salt, and solvent
  • Resolve RNA with aberrant (2' – 5') links from normal SSRNA
Specifications
Specifications
Nonporous
85°C
DNAPac
Ion Exchange
Responsibly packaged
Each
Quaternary Ammonium
Ion Exchange
Bio-LC
Videos
SDS
Documents
Documents
Product Certifications
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Product Content Correction
Product Title
Thermo Scientific™ DNAPac™ PA200 Oligonucleotide HPLC Columns