missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tristel Distel Dosing System, 1 L Bottle of Concentrate
The DISTEL DOSING SYSTEM generates measured doses of QAC-based surface disinfectant, for increased compliance. Designed for the cleaning and disinfection of hard non-porous environmental surfaces.
Brand: Tristel CCH050201
This item is not returnable.
View return policy
Description
- A measured 5 mL dose each time
- Can be used with your choice of spreader, which does not have to be plastic
- Effective against bacteria, yeast and viruses
- Destroys DNA and RNA
- A 1 L bottle generates 2000 fresh doses of surface disinfectant
Specifications
Dosing System | |
Bottle | |
1 Ea. | |
1 L |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Tristel Distel Dosing System, 1 L Bottle of Concentrate > 1 L Bottle of Concentrate
Spot an opportunity for improvement?Share a Content Correction