Learn More
DLin-KC2-DMA, MedChemExpress
MedChemExpress DLin-KC2-DMA is a cationic/ionizable lipid for siRNA delivery.
Brand: Medchem Express HY-112758
Description
DLin-KC2-DMA is a cationic/ionizable lipid for siRNA delivery.Specifications
DLin-KC2-DMA | |
Colorless | |
C43H79NO2 | |
Ethanol : 100 mg/mL (155.74 mM; Need ultrasonic) ∣DMSO : 100 mg/mL (155.74 mM; Need ultrasonic) | |
642.09 | |
98.0% | |
Liquid |
1190197-97-7 | |
Research | |
10 mg | |
CCCCC/C=C\C/C=C\CCCCCCCCC1(CCCCCCCC/C=C\C/C=C\CCCCC)OC(CCN(C)C)CO1 | |
642.09 | |
Research |