missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ DMSO, Anhydrous

DMSO, Anhydrous

Brand:  Molecular Probes™ D12345

88.92 GBP valid until 2025-04-18
Use promo code "24107" to get your promotional price.


Product Code. 13406448

  • £117.00 / Pack of 10

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Dimethyl sulfoxide, or DMSO, is a highly polar organic solvent that is miscible with water. Many compounds that are not directly soluble in water can be dissolved with DMSO to create a concentrated stock solution that can be further diluted to the desired final concentration in aqueous buffer or media at the time of use.

Molecular Probes™ DMSO is suitable for preparing stock solutions of many popular fluorescent reagents including:
  • Fluorescent calcium indicators (e.g. Fluo-3, AM, Fura-2, AM)
  • Fluorescent probes for labeling mitochondria (e.g. rhodamine 123, JC-1, MitoTracker™ Green or Red)
  • Fluorescent probes for cell tracking or tracing (e.g. CFSE, calcein, AM, CellTracker™ Green CMFDA)
  • Fluorescent probes for labeling membranes (e.g. DiI or DiO)
  • Fluorescent reactive oxygen species (ROS) indicators (e.g. DCFDA, CM-DCFDA, dihydroethidium or Amplex Red™ reagent)
  • Amine-reactive fluorescent labeling reagents (e.g. Alexa Fluor™ 488 NHS ester or fluorescein isothiocyanate (FITC))
  • Thiol-reactive fluorescent labeling reagents (e.g. Alexa Fluor™ 488 or fluorescein maleimide)

    Molecular Probes™ DMSO is provided in convenient single-use aliquots to ensure fresh, high quality DMSO every time you prepare a concentrated DMSO stock solution. Please refer to the individual online product manuals for specific instructions on the recommended DMSO stock and working solution concentration for your product.

    Use our Daily Calcs Science Calculator app to simplify making stock solutions and dilution calculations.
    TRUSTED_SUSTAINABILITY
  • Specifications

    Specifications

    6 to 8
    188°C to 190°C
    61 to 64% (Upper), 2.4 to 2.8% (Lower)
    Miscible
    18°C to 19°C
    10 x 3 mL
    87 to 89°C
    Miscible in water
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Manuals

    Certificates

    Certificates

    A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

    5 results found
    Lot Number Certificate Type Date Catalog Number
    Lot Number241024023 Certificate TypeCertificate of Analysis Date13/11/2024 Catalog NumberD12345
    Lot Number241009005 Certificate TypeCertificate of Analysis Date07/11/2024 Catalog NumberD12345
    Lot Number240925001 Certificate TypeCertificate of Analysis Date09/10/2024 Catalog NumberD12345
    Lot Number240819001 Certificate TypeCertificate of Analysis Date13/09/2024 Catalog NumberD12345
    Lot Number240712011 Certificate TypeCertificate of Analysis Date13/08/2024 Catalog NumberD12345
    Special Offers

    Special Offers

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title
    Molecular Probes™ DMSO, Anhydrous > 10 x 3mL

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

    For Research Use Only. Not for use in diagnostic procedures.