missing translation for 'onlineSavingsMsg'
Learn More
Drierite™, without indicator, 8 mesh, Thermo Scientific Chemicals Product Code.: 10462411

Drierite™, without indicator, 8 mesh, Thermo Scientific Chemicals

Product Code. 10462411
500 g, Glass Bottle
Click to view available options
Quantity:
500 g
2 kg
10 kg
Packaging:
Glass Bottle
Plastic Bottle
Plastic Drum
This item is not returnable. View return policy

Product Code. 10462411

Brand: Thermo Scientific Acros 219055000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Adsorbent, Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7778-18-9
Molecular Formula CaO4S
Molecular Weight (g/mol) 136.13
MDL Number MFCD00010912
InChI Key OSGAYBCDTDRGGQ-UHFFFAOYSA-L
PubChem CID 24497
ChEBI CHEBI:31346
SMILES [Ca++].[O-]S([O-])(=O)=O

Specifications

Melting Point 1450.0°C
Physical Form Granular
Quantity 500 g
Packaging Glass Bottle
Solubility Information Solubility in water: ca. 0.25g/L (20°C). Other solubilities: insoluble in organic liquids, slightly soluble in glycerol
IUPAC Name calcium;sulfate
Formula Weight 136.14
Chemical Name or Material Drierite™

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0469054 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number219050100,219050020,219055000
Lot NumberA0469055 Certificate TypeCertificate of Analysis Date13/01/2025 Catalog Number219050100,219050020,219055000
Lot NumberA0464644 Certificate TypeCertificate of Analysis Date23/07/2024 Catalog Number219050100,219050020,219055000
Lot NumberA0464643 Certificate TypeCertificate of Analysis Date23/07/2024 Catalog Number219050100,219050020,219055000
Lot NumberA0449338 Certificate TypeCertificate of Analysis Date12/01/2024 Catalog Number219050100,219050020,219055000
Product Identifier
  • Drierite]TS, without indicator

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Drierite™, without indicator, 8 mesh, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.